missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene, 96%

Product Code. 11343826
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11343826 25 g 25g
11333826 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343826 Supplier Thermo Scientific Alfa Aesar Supplier No. L16894.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 802-93-7 | C12H6F12O2 | 410.159 g/mol

1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene is used to determine the specific details of electrical magnitudes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene is used to determine the specific details of electrical magnitudes.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 802-93-7
Molecular Formula C12H6F12O2
Molecular Weight (g/mol) 410.159
MDL Number MFCD00042090
InChI Key PGUIOHNOYADLMU-UHFFFAOYSA-N
Synonym 1,3-bis 2-hydroxyhexafluoroisopropyl benzene, 1,3-bis hexafluoro-2-hydroxyisopropyl benzene, 1,3-bis alpha-hydroxyhexafluoroisopropyl benzene, alpha,alpha,alpha',alpha'-tetrakis trifluoromethyl-1,3-benzenedimethanol, a,a,a',a'-tetrakis trifluoromethyl-1,3-benzenedimethanol, 1,3-benzenedimethanol, .alpha.,.alpha.,.alpha.',.alpha.'-tetrakis trifluoromethyl, 1,3-bis hexafluoro-alpha-hydroxyisopropyl benzene, 1,1,1,3,3,3-hexafluoro-2-3-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl propan-2-ol, 1,3-bis 1,1,1,3,3,3-hexafluoro-2-hydroxypropyl benzene, 1,3 hfab
PubChem CID 69933
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-[3-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol
SMILES C1=CC(=CC(=C1)C(C(F)(F)F)(C(F)(F)F)O)C(C(F)(F)F)(C(F)(F)F)O

Tekniske data

Melting Point 9°C to 10°C
Density 1.659
Boiling Point 99°C to 100°C (20 mmHg)
Flash Point 97°C (206°F)
Refractive Index 1.389
Quantity 25 g
Beilstein 2024879
Solubility Information Difficult to mix.
Formula Weight 410.16
Percent Purity 96%
Chemical Name or Material 1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.