missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(4-bromophenyl)-5-phenyl-2,4-imidazolidinedione, Thermo Scientific™

Product Code. 15456179
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456179 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15456179 Supplier Thermo Scientific Alfa Aesar Supplier No. J64065.MF
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

An imidazolidine-2,-4-dione derivative that is an effective and selective inverse agonist of CB cannabinoid receptor.
TRUSTED_SUSTAINABILITY

Specifications

CAS 878533-35-8
Color White
Molecular Formula C21H14Br2N2O2
MDL Number MFCD12912318
Quantity 50 mg
Synonym 1,3-bis 4-bromophenyl-5-phenyl-2,4-imidazolidinedione, 1,3-bis 4-bromophenyl-5-phenylimidazolidine-2,4-dione
InChI Key ROASQEPZAKGACK-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2C(=O)N(C(=O)N2C3=CC=C(C=C3)Br)C4=CC=C(C=C4)Br
IUPAC Name 1,3-bis(4-bromophenyl)-5-phenylimidazolidine-2,4-dione
Molecular Weight (g/mol) 486.163
PubChem CID 11851013
Formula Weight 486.16
Physical Form Solid
Chemical Name or Material 1,3-Bis(4-bromophenyl)-5-phenyl-2,4-imidazolidinedione
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.