missing translation for 'onlineSavingsMsg'
Learn More

[1,3-Bis(diphenylphosphino)propane]nickel(II) chloride, 98+%

Product Code. 10618134 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10618134 25 g Glass bottle 25g
10486573 1 g Glass bottle 1g
10452251 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10618134 Supplier Thermo Scientific Supplier No. 291590250
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15629-92-2 | C27H26Cl2NiP2 | 542.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15629-92-2
Molecular Formula C27H26Cl2NiP2
Molecular Weight (g/mol) 542.04
MDL Number MFCD00015318
InChI Key REQVOKPBMSCTNI-UHFFFAOYSA-L
PubChem CID 131675641
IUPAC Name 3-diphenylphosphanylpropyl(diphenyl)phosphane;nickel;dihydrochloride
SMILES Cl[Ni++]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 213.0°C
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Fieser 06,50; 15,122
Solubility Information Solubility in water: insoluble
Formula Weight 542.04
Percent Purity 99%
Physical Form Crystals or Powder
Chemical Name or Material [1,3-Bis(diphenylphosphino)propane]nickel(II) chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • [1,3-Bis(diphenylphosphino)propane]nickel(II) chloride
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H350-May cause cancer.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.