missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis[(trimethylsilyl)ethynyl]benzene, 96%, Alfa Aesar™

Product Code. 11312926
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312926 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11312926 Supplier Thermo Scientific Alfa Aesar Supplier No. L10640.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 38170-80-8
Molecular Formula C16H22Si2
Molecular Weight (g/mol) 270.522
MDL Number MFCD00078305
InChI Key ASZOZZMGMNVKDV-UHFFFAOYSA-N
Synonym 1,3-bis trimethylsilyl ethynyl benzene, 1,3-bis trimethylsilylethynyl benzene, trimethyl 2-3-2-trimethylsilyl ethynyl phenyl ethynyl silane, acmc-209iy7, 1,3-bis trimethylsilyl ethynylbenzene, 1,3-bis 2-trimethylsilyl ethynyl benzene, 1,3-bis-trimethylsilyl-ethynyl-benzene, benzene,1,3-bis 2-trimethylsilyl ethynyl, trimethyl-2-3-2-trimethylsilylethynyl phenyl ethynyl silane
PubChem CID 142239
IUPAC Name trimethyl-[2-[3-(2-trimethylsilylethynyl)phenyl]ethynyl]silane
SMILES C[Si](C)(C)C#CC1=CC(=CC=C1)C#C[Si](C)(C)C

Specifications

Melting Point 57°C to 59°C
Quantity 5 g
Beilstein 2845239
Formula Weight 270.53
Percent Purity 96%
Chemical Name or Material 1,3-Bis[(trimethylsilyl)ethynyl]benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.