missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis[tris(hydroxymethyl)amino]propane, 99%

Product Code. 15189223 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
1kg
250g
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15189223 1 kg Plastic Bottle 1kg
10516440 25 g Glass bottle 25g
10526440 100 g Glass bottle 100g
10535480 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15189223 Supplier Thermo Scientific Acros Supplier No. 202640010
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 64431-96-5 | C11H26N2O6 | 282.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64431-96-5
Molecular Formula C11H26N2O6
Molecular Weight (g/mol) 282.33
MDL Number MFCD00004689
InChI Key HHKZCCWKTZRCCL-UHFFFAOYSA-N
Synonym bis-tris propane, 1,3-bis tris hydroxymethyl methylamino propane, 2,2'-propane-1,3-diylbis azanediyl bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-dihydroxymethyl-ethylamino-propylamino-2-hydroxymethyl-propane-1,3-diol, 1,3-propanediol, 2,2'-1,3-propanediyldiimino bis 2-hydroxymethyl, 2,2'-propane-1,3-diyldiimino bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino propylamino-2-hydroxymethyl propane-1,3-diol, b3p
PubChem CID 125132
ChEBI CHEBI:40947
IUPAC Name 2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CNC(CO)(CO)CO)CNC(CO)(CO)CO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 164.0°C to 165.0°C
Color White
Elemental Analysis 46.33 to 47.2% C, 9.82 to 10.02% N
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic Bottle
Linear Formula CH2{CH2NHC(CH2OH)3]2
Quantity 1 kg
Beilstein 04,III,859
Solubility Information Solubility in water: soluble in cold water
Formula Weight 282.33
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis[tris(hydroxymethyl)amino]propane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.