missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,3-Bis[tris(hydroxymethyl)methylamino]propane, 98+%

Product Code. 11305837
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11305837 25 g 25g
11315837 100 g 100g
11325837 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11305837 Supplier Thermo Scientific Chemicals Supplier No. 043496.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 64431-96-5 | C11H26N2O6 | 282.337 g/mol

1,3-Bis[tris(hydroxymethyl)methylamino]propane is used as a buffer for polymerase chain reaction and molecular biology. It is also used with hydrochloric acid buffer for stabilization of farnesyl diphosphate isolated from a strain of saccharomyces cerevisiae. It acts as a ligand and forms dinuclear hydroxo complexes with Lanthanides(III). Further, it is useful for calibration of genetically encoded pH indicators expressed in the cytosol or mitochondria. It is also employed to investigate the influence of anions with varying nucleophilic properties on the autoxidation of oxymyoglobin. In addition to this, it is used in the purification of glucose-binding protein from membranes of sulfolobus solfataricus and as a non-zwitterionic buffer, which is used in the pH range between 6.3 and 9.5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64431-96-5
Molecular Formula C11H26N2O6
Molecular Weight (g/mol) 282.337
MDL Number MFCD00004689
InChI Key HHKZCCWKTZRCCL-UHFFFAOYSA-N
Synonym bis-tris propane, 1,3-bis tris hydroxymethyl methylamino propane, 2,2'-propane-1,3-diylbis azanediyl bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-dihydroxymethyl-ethylamino-propylamino-2-hydroxymethyl-propane-1,3-diol, 1,3-propanediol, 2,2'-1,3-propanediyldiimino bis 2-hydroxymethyl, 2,2'-propane-1,3-diyldiimino bis 2-hydroxymethyl propane-1,3-diol, 2-3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino propylamino-2-hydroxymethyl propane-1,3-diol, b3p
PubChem CID 125132
ChEBI CHEBI:40947
IUPAC Name 2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CNC(CO)(CO)CO)CNC(CO)(CO)CO

Specifications

Melting Point 168°C to 170°C
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 282.34
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material 1,3-Bis[tris (hydroxymethyl)methylamino]propane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.