missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Bromophenyl)piperidine, ≥97%, Thermo Scientific™

Product Code. 10025852
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10025852 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10025852 Supplier Thermo Scientific Maybridge Supplier No. CC34010DA
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 84964-24-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H14BrN
MDL Number MFCD04108532
Quantity 1 g
Synonym 1-3-bromophenyl piperidine, 3-bromophenyl piperidine, acmc-209pz4, n-3-bromophenyl piperidine, 1-piperidino-3-bromobenzene
InChI Key PMTXPOLLPCSCBE-UHFFFAOYSA-N
SMILES BrC1=CC=CC(=C1)N1CCCCC1
IUPAC Name 1-(3-bromophenyl)piperidine
Molecular Weight (g/mol) 240.14
PubChem CID 4961269
Formula Weight 240.14
Percent Purity 97%
Chemical Name or Material 1-(3-Bromophenyl)piperidine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.