missing translation for 'onlineSavingsMsg'
Learn More

1-[3-chloro-5-(trifluoromethyl)-2-pyridyl]piperazine, 97%, Thermo Scientific™

Product Code. 10753663
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10753663 1 g Amber Glass Bottle 1g
10190652 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10753663 Supplier Thermo Scientific Maybridge Supplier No. SEW04036DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 132834-59-4
Molecular Formula C10H11ClF3N3
Molecular Weight (g/mol) 265.66
MDL Number MFCD00173934
InChI Key RRIPRMGRQRYRRG-UHFFFAOYSA-N
Synonym 1-3-chloro-5-trifluoromethyl-2-pyridyl piperazine, 1-3-chloro-5-trifluoromethyl pyridin-2-yl piperazine, 1-3-chloro-5-trifluoromethyl pyrid-2-yl piperazine, 1-3-chloro-5-trifluoromethyl-2-pyridinyl piperazine, piperazine, 1-3-chloro-5-trifluoromethyl-2-pyridinyl, 3-chloro-5-trifluoromethyl-2-piperazinopyridine, 1-3-chloro-5-trifluoromethyl-pyridin-2-yl-piperazine, piperazine,1-3-chloro-5-trifluoromethyl-2-pyridinyl
PubChem CID 667619
IUPAC Name 1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperazine
SMILES FC(F)(F)C1=CC(Cl)=C(N=C1)N1CCNCC1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 265.66
Percent Purity 97%
Chemical Name or Material 1-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.