missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Chlorophenyl)biguanide hydrochloride, 97%

Product Code. 11464247
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464247 1 g 1g
11474247 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464247 Supplier Thermo Scientific Alfa Aesar Supplier No. B20355.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2113-05-5 | C8H11Cl2N5 | 248.11 g/mol

Specific agonist for 5-HT receptor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2113-05-5
MDL Number MFCD00053019

Specifications

Melting Point 191°C to 194°C
Molecular Formula C8H11Cl2N5
Quantity 1 g
Synonym 1-3-chlorophenyl biguanide hydrochloride, m-chlorophenylbiguanide hydrochloride, 1-m-chlorophenyl-biguanide hydrochloride, m-cpbg hydrochloride, 1-m-chlorophenyl biguanide hydrochloride, 1-carbamimidamido-n-3-chlorophenyl methanimidamide hydrochloride, imidodicarbonimidic diamide, n-3-chlorophenyl-, monohydrochloride, n-m-chlorophenylbiguanide hydrochloride, biguanide, 1-m-chlorophenyl-, monohydrochloride, n-3-chlorophenyl imidodicarbonimidic diamide hydrochloride
InChI Key FOWAIJYHRWFTHR-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)Cl)N=C(N)N=C(N)N.Cl
IUPAC Name 2-(3-chlorophenyl)-1-(diaminomethylidene)guanidine;hydrochloride
Molecular Weight (g/mol) 248.11
PubChem CID 2730228
Formula Weight 248.12
Percent Purity 97%
Chemical Name or Material 1-(3-Chlorophenyl)biguanide hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.