missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Chlorophenyl)biguanide hydrochloride, 97%

Product Code. 11474247
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474247 5 g 5g
11464247 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474247 Supplier Thermo Scientific Alfa Aesar Supplier No. B20355.06
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2113-05-5 | C8H11Cl2N5 | 248.11 g/mol

Specific agonist for 5-HT receptor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2113-05-5
MDL Number MFCD00053019

Specifications

Melting Point 191°C to 194°C
Molecular Formula C2H6N4S
Quantity 5 g
Sensitivity Hygroscopic
Synonym amidinothiourea, guanylthiourea, guthimin, guthimine, 2-imino-4-thiobiuret, n-amidinothiourea, thiourea, aminoiminomethyl, thiodicyanodiamidine, diaminomethylidene thiourea, gutimine
InChI Key OKGXJRGLYVRVNE-UHFFFAOYSA-N
SMILES C(=NC(=S)N)(N)N
IUPAC Name diaminomethylidenethiourea
Molecular Weight (g/mol) 118.16
PubChem CID 2724563
Formula Weight 248.12
Percent Purity 97%
Chemical Name or Material 1-(3-Chlorophenyl)biguanide hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.