missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Chlorophenyl)piperazine Hydrochloride, TRC

Product Code. 30364516
Change view
Click to view available options
Quantity:
10 mg
50 mg
10 g
Unit Size:
10g
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364516 10 g 10g
30349272 10 mg 10mg
30351410 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364516 Supplier Toronto Research Chemicals Supplier No. TRCC37959110G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13078-15-4
Molecular Formula C10 H13 Cl N2 . Cl H
Molecular Weight (g/mol) 233.14
Synonym 1-(3-Chlorophenyl)piperazine Monohydrochloride, 1-(m-Chlorophenyl)piperazine Monohydrochloride, 1-(3-Chlorophenyl)piperazine Hydrochloride, 1-(m-Chlorophenyl)piperazine hydrochloride, N-(3-Chlorophenyl)piperazine hydrochloride, m-CPP hydrochloride
IUPAC Name 1-(3-chlorophenyl)piperazine;hydrochloride
SMILES Cl.Clc1cccc(c1)N2CCNCC2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H13ClN2.ClH/c11-9-2-1-3-10(8-9)13-6-4-12-5-7-13;/h1-3,8,12H,4-7H2;1H
Formula Weight 232.0534
Chemical Name or Material 1-(3-Chlorophenyl) piperazine (hydrochloride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.