missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cycloheptanedione, 97%

Product Code. 15460848
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460848 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15460848 Supplier Thermo Scientific Alfa Aesar Supplier No. H54588.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1194-18-9 | C7H10O2 | 126.155 g/mol

1,3-Cyclopentanedione was used in the synthesis of chemical probes for selective labeling of sulfuric acid proteins. is a versatile reagent for synthesis of perhydroazulenes, prostaglandin products (biologically active unsaturated 20-carbon fatty acids including a 5-carbon ring, derived enzymatically ), and Knoevenagel products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Cyclopentanedione was used in the synthesis of chemical probes for selective labeling of sulfuric acid proteins. is a versatile reagent for synthesis of perhydroazulenes, prostaglandin products (biologically active unsaturated 2
TRUSTED_SUSTAINABILITY

Specifications

CAS 1194-18-9
Density 1.1
Boiling Point 254°C
Flash Point >110°C (230°F)
Molecular Formula C7H10O2
Refractive Index 1.483
MDL Number MFCD01863735
Quantity 1 g
Synonym 1,3-cycloheptanedione, 1 3-cycloheptanedione 97, acmc-20aijy, 1,3-cycloheptandione, pubchem20261, cycloheptan-1,3-dione, cyclohep tane-1,3-dione, cycloheptane-1,3-quinone, cycloheptane-?1,?3-?dione, ksc173k8n
Solubility Information Soluble in water (5.5g/100 g),and dimethyl formamide (DMF)
InChI Key DBOVMTXPZWVYAQ-UHFFFAOYSA-N
SMILES C1CCC(=O)CC(=O)C1
IUPAC Name cycloheptane-1,3-dione
Molecular Weight (g/mol) 126.155
PubChem CID 4072367
Formula Weight 126.15
Percent Purity 97%
Chemical Name or Material 1,3-Cycloheptanedione
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.