missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diacetylbenzene, 99%, Thermo Scientific Chemicals

Product Code. 10347970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10347970 5 g Glass bottle 5g
10337970 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10347970 Supplier Thermo Scientific Acros Supplier No. 170090050
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6781-42-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00008740
InChI Key VCHOFVSNWYPAEF-UHFFFAOYSA-N
Synonym 1,3-diacetylbenzene, 1,1'-1,3-phenylene diethanone, ethanone, 1,1'-1,3-phenylene bis, m-diacetylbenzene, benzene-1,3-bis acetyl, m-acetylacetophenone, m-acetyl acetophenone, 1-3-acetylphenyl ethanone, 1-3-acetylphenyl ethan-1-one, m-diacetyl benzene
PubChem CID 23229
IUPAC Name 1-(3-acetylphenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)C(C)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 35.0°C to 37.0°C
Color Yellow
Boiling Point 150.0°C to 155.0°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(COCH3)2
Refractive Index 1.5485 to 1.5505
Quantity 5 g
Formula Weight 162.19
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material 1, 3-Diacetylbenzene, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.