missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibenzyl-1H-benzo[d]imidazol-3-ium Chloride, TRC

Product Code. 30357807
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357807 500 mg 500mg
30367355 100 mg 100mg
30359580 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357807 Supplier Toronto Research Chemicals Supplier No. TRCD159141500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36339-13-6
Molecular Formula C21H19ClN2
Molecular Weight (g/mol) 334.85
Synonym 1,3-Dibenzyl-1H-1,3-benzodiazol-3-ium Chloride, 1,3-Dibenzylbenzimidazolium Chloride 1,3-Dibenzylbenzimidzolium Chloride, N,N'-Dibenzylbenzimidazolium Chloride
IUPAC Name 1,3-dibenzylbenzimidazol-3-ium;chloride
SMILES [Cl-].C(c1ccccc1)[n+]2cn(Cc3ccccc3)c4ccccc24

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C21H19N2.ClH/c1-3-9-18(10-4-1)15-22-17-23(16-19-11-5-2-6-12-19)21-14-8-7-13-20(21)22;/h1-14,17H,15-16H2;1H/q+1;/p-1
Formula Weight 334.1237
Chemical Name or Material 1,3-Dibenzyl-1H-benzo[d]imidazol-3-ium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.