missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-2-propanol, 95%

Product Code. 10569082 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10569082 100 g Glass bottle 100g
10055882 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10569082 Supplier Thermo Scientific Acros Supplier No. 406541000
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 96-21-9 | C3H6Br2O | 217.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-21-9
Molecular Formula C3H6Br2O
Molecular Weight (g/mol) 217.89
MDL Number MFCD00000216
InChI Key KIHQZLPHVZKELA-UHFFFAOYSA-N
Synonym 1,3-dibromo-2-propanol, 2-propanol, 1,3-dibromo, 1,3-dibromohydrin, glycerol 1,3-dibromohydrin, 2-hydroxy-1,3-dibromopropane, 1,3-dibromo-2-hydroxypropane, alpha-dibromohydrin, .alpha.-dibromohydrin, glycerol alpha,gamma-dibromohydrin, glycerol alpha,gamma-dibromohydrine
PubChem CID 7287
IUPAC Name 1,3-dibromopropan-2-ol
SMILES OC(CBr)CBr

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Brown or Colorless to Yellow
Density 2.1300g/mL
Boiling Point 219°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH2BrCHOHCH2Br
Refractive Index 1.5500 to 1.554
Quantity 100 g
Beilstein 01, 365
Specific Gravity 2.13
Solubility Information Solubility in water: soluble. Other solubilities: very soluble in acetone,ethyl ether and ethanol
Formula Weight 217.89
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1, 3-Dibromo-2-propanol, pract., 85%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,3-Dibromo-2-propanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.