missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-5,5-dimethylhydantoin, 98%

Product Code. 11422917
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422917 2500 g 2500g
11402917 100 g 100g
11412917 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11422917 Supplier Thermo Scientific Alfa Aesar Supplier No. A15510.0E
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 77-48-5 | C5H6Br2N2O2 | 285.923 g/mol

It is used for drinking water purification, recreational water treatment, as a bleaching agent in pulp and paper mills, and for treating industrial/commercial water cooling systems. It is an efficient and selective agent for the oxidation of thiols to disulfides in solution or under solvent-free conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for drinking water purification, recreational water treatment, as a bleaching agent in pulp and paper mills, and for treating industrial/commercial water cooling systems. It is an efficient and selective agent for the oxidation of thiols to disulfides in solution or under solvent-free conditions.

Solubility
Soluble in Alcohol, Ether, Benzene. Insoluble in Water.

Notes
Moisture sensitive. Store away from oxidizing agents, reducing agents, flammable substances, organic materials, metal powders & moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-48-5
Molecular Formula C5H6Br2N2O2
Molecular Weight (g/mol) 285.923
MDL Number MFCD00003189
InChI Key VRLDVERQJMEPIF-UHFFFAOYSA-N
Synonym 1,3-dibromo-5,5-dimethylhydantoin, dibromantin, dibromantine, take charge orange, dbdmh, n,n'-dibromodimethylhydantoin, 5,5-dimethyl-1,3-dibromohydantoin, 2,4-imidazolidinedione, 1,3-dibromo-5,5-dimethyl, 1,3-dibromo-5,5-dimethyl-2,4-imidazolidinedione, unii-v9r5f9i7mz
PubChem CID 6479
IUPAC Name 1,3-dibromo-5,5-dimethylimidazolidine-2,4-dione
SMILES CC1(C(=O)N(C(=O)N1Br)Br)C

Specifications

Melting Point ∼198°C (decomposition)
Flash Point 155°C (311°F)
Quantity 2500 g
UN Number UN3085
Beilstein 146024
Sensitivity Moisture sensitive
Solubility Information Soluble in Alcohol,Ether,Benzene. Insoluble in Water.
Formula Weight 285.93
Percent Purity 98%
Chemical Name or Material 1,3-Dibromo-5,5-dimethylhydantoin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.