missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromopropane, 98%, stabilized

Product Code. 10367470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10367470 100 mL Glass bottle 100mL
10563701 500 mL Glass bottle 500mL
10155353 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10367470 Supplier Thermo Scientific Acros Supplier No. 155041000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 109-64-8 | C3H6Br2 | 201.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-64-8
Molecular Formula C3H6Br2
Molecular Weight (g/mol) 201.89
MDL Number MFCD00000255
InChI Key VEFLKXRACNJHOV-UHFFFAOYSA-N
Synonym trimethylene bromide, trimethylene dibromide, propane, 1,3-dibromo, alpha,gamma-dibromopropane, unii-yqr3048ix9, ccris 6711, .alpha.,.gamma.-dibromopropane, 1,3-dibromo propane, 1,3-dibromo-propane, .omega.,.omega.'-dibromopropane
PubChem CID 8001
IUPAC Name 1,3-dibromopropane
SMILES BrCCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -34.0°C
Color Colorless to Yellow
Density 1.9890g/mL
Boiling Point 167.0°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Br(CH2)3Br
Refractive Index 1.5215 to 1.5235
Quantity 100 mL
Beilstein 01, 110
Fieser 13,313
Merck Index 15, 9884
Specific Gravity 1.989
Solubility Information Solubility in water: 1.7g/l (20°C). Other solubilities: soluble in organic solvents
Stabilizer 400ppm butylene oxide
Formula Weight 201.89
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,3-Dibromopropane, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,3-Dibromopropane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.