missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromopropane, TRC

Product Code. 30384101
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384101 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384101 Supplier Toronto Research Chemicals Supplier No. TRCD425850500MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material 1,3-Dibromopropane
CAS 109-64-8
Molecular Formula C3 H6 Br2
Quantity 500 mg
Synonym Propane, 1,3-dibromo- (8CI, 9CI, ACI), 1,3-Dibromopropane (ACI), 1,3-Bis(bromo)propane, NSC 62663, Trimethylene bromide, Trimethylene dibromide, α,γ-Dibromopropane, ω,ω'-Dibromopropane
InChI Formula InChI=1S/C3H6Br2/c4-2-1-3-5/h1-3H2
SMILES BrCCCBr
IUPAC Name 1,3-dibromopropane
Molecular Weight (g/mol) 201.89
Formula Weight 199.8836
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.