missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dicaffeoylquinic Acid, TRC

Product Code. 30134755
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134755 25 mg 25mg
30007798 2.5 mg 2.5mg
30010931 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134755 Supplier Toronto Research Chemicals Supplier No. TRCD42970025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19870-46-3
Molecular Formula C25 H24 O12
Molecular Weight (g/mol) 516.45
Synonym Cyclohexanecarboxylic acid, 1,3-bis[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]-4,5-dihydroxy-, (1S,3R,4R,5R)-, Cinnamic acid, 3,4-dihydroxy-, 1-carboxy-4,5-dihydroxy-1,3-cyclohexylene ester (8CI), Cyclohexanecarboxylic acid, 1,3-bis[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-4,5-dihydroxy-, (1S,3R,4R,5R)- (9CI), Cyclohexanecarboxylic acid, 1,3-bis[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-4,5-dihydroxy-, [1S-(1α,3β,4α,5α)]-, (1S,3R,4R,5R)-1,3-Bis[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]-4,5-dihydroxycyclohexanecarboxylic acid, 1,3-Dicaffeoylquinic acid, 1,3-O-Dicaffeoylquinic acid, 1,5-Dicaffeoylquinic acid
IUPAC Name (1S,3R,4R,5R)-1,3-bis[[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy]-4,5-dihydroxycyclohexane-1-carboxylic acid
SMILES O[C@@H]1C[C@@](C[C@@H](OC(=O)\C=C\c2ccc(O)c(O)c2)[C@@H]1O)(OC(=O)\C=C\c3ccc(O)c(O)c3)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C25H24O12/c26-15-5-1-13(9-17(15)28)3-7-21(31)36-20-12-25(24(34)35,11-19(30)23(20)33)37-22(32)8-4-14-2-6-16(27)18(29)10-14/h1-10,19-20,23,26-30,33H,11-12H2,(H,34,35)/b7-3+,8-4+/t19-,20-,23-,25+/m1/s1
Formula Weight 516.1268
Chemical Name or Material 1,3-Dicaffeoylquinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.