Learn More
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane, 97%
Description
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane is a silylating reagent used for simultaneous protection of 3'- and 5'-hydroxy functions of ribonucleosides. It is used in the analysis of Watson-Crick and Hoogstein base pairing in nucleotides. It is also employed in the formation of ribavirin chemical delivery systems. Further, it serves as a protecting group reagent for open-chain polyhydroxy compounds. In addition to this, it is involved in the preparation of cyclic bridged peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane is a silylating reagent used for simultaneous protection of 3′- and 5′-hydroxy functions of ribonucleosides. It is used in the analysis of Watson-Crick and Hoogstein base pairing in nucleotides. It is also employed in the formation of ribavirin chemical delivery systems. Further, it serves as a protecting group reagent for open-chain polyhydroxy compounds. In addition to this, it is involved in the preparation of cyclic bridged peptides.
Solubility
Miscible with dimethylformamide, hexane, ethyl acetate, chloroform and dichloromethane.
Notes
Incompatible with strong oxidizing agents and water.
Specifications
Specifications
| Boiling Point | 69°C to 70°C (0.5mmHg) |
| Quantity | 1 g |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.454 |
| Sensitivity | Moisture sensitive |
| Density | 0.986 |
| UN Number | UN1760 |
| Beilstein | 1934216 |
| Solubility Information | Miscible with dimethylformamide,hexane,ethyl acetate,chloroform and dichloromethane. |
| Formula Weight | 315.43 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.