missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-2-propanol, 99%

Product Code. 10184290 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10184290 1 L Glass bottle 1 L
10032320 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10184290 Supplier Thermo Scientific Acros Supplier No. 180180010
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 96-23-1 | C3H6Cl2O | 128.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-23-1
Molecular Formula C3H6Cl2O
Molecular Weight (g/mol) 128.98
MDL Number MFCD00000951
InChI Key DEWLEGDTCGBNGU-UHFFFAOYSA-N
Synonym 1,3-dichloro-2-propanol, dichlorohydrin, 1,3-dichloroisopropanol, 1,3-dichlorohydrin, glycerol 1,3-dichlorohydrin, 2-propanol, 1,3-dichloro, enodrin, 1,3-dichloro-2-hydroxypropane, 1,3-dichloroisopropyl alcohol, sym-glycerol dichlorohydrin
PubChem CID 7289
ChEBI CHEBI:18917
IUPAC Name 1,3-dichloropropan-2-ol
SMILES OC(CCl)CCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -4.0°C
Color Colorless to Yellow
Density 1.3510g/mL
Boiling Point 172.0°C to 176.0°C
Flash Point 74°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CH(OH)CH2Cl
Refractive Index 1.4820 to 1.4840
Quantity 1 L
Beilstein 01, 364
Merck Index 15, 3084
Specific Gravity 1.351
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: miscible with most common organic solvents
Viscosity 13.9 mPa.s (20°C)
Formula Weight 128.99
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 3-Dichloro-2-propanol, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,3-Dichloro-2-propanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.