missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-5,5-dimethylhydantoin, 98%

Product Code. 11420653
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420653 500 g 500g
11410653 100 g 100g
11430653 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420653 Supplier Thermo Scientific Alfa Aesar Supplier No. A12718.36
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 118-52-5 | C5H6Cl2N2O2 | 197.015 g/mol

Dantoin(R) DCDMH is an industrial bleaching agent based on dimethyl hydantoin. This product finds application wherever solid bleach is desired. It is used as a mild chlorinating agent in organic synthesis.It is also used as a sanitizer in swimming pools, laundries restaurants, cutting oils and pharmaceutical industry as a general cleaner. Other uses include it is been used as a preventer and catalyst in polymer industry, used to emboss or texturize resinous sheet preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dantoin(R) DCDMH is an industrial bleaching agent based on dimethyl hydantoin. This product finds application wherever solid bleach is desired. It is used as a mild chlorinating agent in organic synthesis.It is also used as a sanitizer in swimming pools, laundries restaurants, cutting oils and pharmaceutical industry as a general cleaner. Other uses include it is been used as a preventer and catalyst in polymer industry, used to emboss or texturize resinous sheet preparations.

Solubility
Soluble in water 0.21 g/100 mL (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with reducing agents, acids, strong bases, organic materials, powdered metals. Moisture, light, air sensitive. Stable, but a strong oxidizer.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-52-5
Molecular Formula C5H6Cl2N2O2
Molecular Weight (g/mol) 197.015
MDL Number MFCD00003190
InChI Key KEQGZUUPPQEDPF-UHFFFAOYSA-N
Synonym 1,3-dichloro-5,5-dimethylhydantoin, dantoin, dactin, daktin, dichlorantin, omchlor, halane, hydan, hydantoin, dichlorodimethyl, hydan antiseptic
PubChem CID 8360
IUPAC Name 1,3-dichloro-5,5-dimethylimidazolidine-2,4-dione
SMILES CC1(C(=O)N(C(=O)N1Cl)Cl)C

Specifications

Melting Point 130°C to 134°C
Density 1.5
Flash Point 174°C (345°F)
Quantity 500 g
UN Number UN1479
Beilstein 146013
Sensitivity Moisture sensitive
Merck Index 14,3065
Solubility Information Soluble in water 0.21g/100 mL (25°C).
Formula Weight 197.02
Percent Purity 98%
Chemical Name or Material 1,3-Dichloro-5,5-dimethylhydantoin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.