missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-5-isocyanatobenzene, TRC

Product Code. 30368931
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30368931 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368931 Supplier Toronto Research Chemicals Supplier No. TRCD43489510G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 34893-92-0
Molecular Formula C7H3Cl2NO
Quantity 10 g
Synonym 3,5-Dichloro-1-isocyanatobenzene, 3,5-Dichlorophenyl isocyanate
InChI Formula InChI=1S/C7H3Cl2NO/c8-5-1-6(9)3-7(2-5)10-4-11/h1-3H
SMILES Clc1cc(Cl)cc(c1)N=C=O
IUPAC Name 1,3-dichloro-5-isocyanatobenzene
Molecular Weight (g/mol) 188.01
Formula Weight 186.9592
Chemical Name or Material 1,3-Dichloro-5-isocyanatobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.