missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloroacetone, 96%

Product Code. 11429080
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11429080

Brand: Thermo Scientific Alfa Aesar A14149.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 534-07-6 | C3H4Cl2O | 126.96 g/mol

1,3-Dichloroacetone is used in the synthesis of citric acid. It is used as a solvent and as an intermediate in organic synthesis. Further, it serves as an acceptor substrate in the cross-aldol reaction with donor substrates such as acetone, cyclopentanone and cyclohexanone. It is also used in the preparation of an active pharmaceutical ingredient, famotidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dichloroacetone is used in the synthesis of citric acid. It is used as a solvent and as an intermediate in organic synthesis. Further, it serves as an acceptor substrate in the cross-aldol reaction with donor substrates such as acetone, cyclopentanone and cyclohexanone. It is also used in the preparation of an active pharmaceutical ingredient, famotidine.

Solubility
Soluble in water, ethanol and diethyl ether.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents, strong bases and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-07-6
Molecular Formula C3H4Cl2O
Molecular Weight (g/mol) 126.96
MDL Number MFCD00000937
InChI Key SUNMBRGCANLOEG-UHFFFAOYSA-N
Synonym 1,3-dichloroacetone, 1,3-dichloro-2-propanone, 1,3-dichloropropanone, s-dichloroacetone, sym-dichloroacetone, bis chloromethyl ketone, 2-propanone, 1,3-dichloro, acetone, 1,3-dichloro, chloromethyl ketone
PubChem CID 10793
IUPAC Name 1,3-dichloropropan-2-one
SMILES ClCC(=O)CCl

Specifications

Melting Point 40°C to 44°C
Density 1.383
pH 4.2
Boiling Point 172°C to 173°C
Flash Point 95°C (203°F)
Odor Pungent
Refractive Index 1.44
Quantity 500 g
UN Number UN2649
Beilstein 605456
Merck Index 14,3052
Solubility Information Soluble in water,ethanol and diethyl ether.
Formula Weight 126.97
Percent Purity 96%
Chemical Name or Material 1,3-Dichloroacetone
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H300-Fatal if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.