missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichlorobenzene, TRC

Product Code. 30385255
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
Unit Size:
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385255 250 g -
30378813 500 g 500g
30372947 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385255 Supplier Toronto Research Chemicals Supplier No. TRCD431930250G
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-73-1
Molecular Formula C6 H4 Cl2
Molecular Weight (g/mol) 147
Synonym Benzene, m-dichloro- (8CI), 1,3-Dichlorobenzene, 2,4-Dichlorobenzene, 2,6-Dichlorobenzene, NSC 8754, m-Dichlorobenzene, m-Dichlorobenzol, m-Phenylene dichloride
IUPAC Name 1,3-dichlorobenzene
SMILES Clc1cccc(Cl)c1

Specifications

Quantity 250 g
InChI Formula InChI=1S/C6H4Cl2/c7-5-2-1-3-6(8)4-5/h1-4H
Formula Weight 145.969
Chemical Name or Material 1,3-Dichlorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.