missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloropropane, 95%

Product Code. 11374207
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374207 25 g 25g
11364207 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374207 Supplier Thermo Scientific Alfa Aesar Supplier No. 036685.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 142-28-9 | C3H6Cl2 | 112.981 g/mol

1,3-Dichloropropane is used as an intermediate in organic synthesis. It is also used as solvent, detergent and chromatographic analysis. Further, it is used to prepare 3,7-dithia-nonanedioic acid by reacting with mercaptoacetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-28-9
Molecular Formula C3H6Cl2
Molecular Weight (g/mol) 112.981
MDL Number MFCD00000999
InChI Key YHRUOJUYPBUZOS-UHFFFAOYSA-N
Synonym trimethylene dichloride, propane, 1,3-dichloro, trimethylene chloride, 1,3-dichloro-propane, unii-aj1hq2gucp, ccris 9220, ch2clch2ch2cl, aj1hq2gucp, cl ch2 3cl, 1,3-dichlorpropan
PubChem CID 8881
IUPAC Name 1,3-dichloropropane
SMILES C(CCl)CCl

Specifications

Density 1.190 g/mL
Boiling Point 120°C
Flash Point 20°C (68°F)
Odor Sweet
Refractive Index 1.4481
Quantity 25 g
UN Number UN1993
Solubility Information Miscible with water.
Formula Weight 112.99
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1,3-Dichloropropane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,3-Dichloropropane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 1,3-Dichloropropane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.