missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloropropene, cis + trans, tech. 90%, Thermo Scientific Chemicals

Product Code. 11358375
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11358375 25 g 25g
11368375 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 11358375

Brand: Thermo Scientific Alfa Aesar L10332.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-75-6
Molecular Formula C3H4Cl2
Molecular Weight (g/mol) 110.97
MDL Number MFCD00000985
InChI Key UOORRWUZONOOLO-UPHRSURJSA-N
Synonym 1,3-dichloropropene, trans-1,3-dichloropropene, telone ii, telone, 1,3-dichloropropylene, nemex, trans-1,3-dichloropropylene, dorlone ii, telone c, di-trapex cp
PubChem CID 24726
ChEBI CHEBI:18624
SMILES ClC\C=C/Cl

Specifications

Melting Point ∼-84°C
Boiling Point 105°C to 106°C
Flash Point 21°C (70°F)
Odor Chloroform-like
Quantity 25 g
UN Number UN2047
Beilstein 1719556
Merck Index 14,3075
IUPAC Name (1Z)-1,3-dichloroprop-1-ene
Formula Weight 110.97
Percent Purity 90%
Grade Technical
Chemical Name or Material 1,3-Dichloropropene, Cis + Trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.