missing translation for 'onlineSavingsMsg'
Learn More

1,3-Difluoro-2-nitrobenzene, 98%

Product Code. 15412018
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15412018 1 g 1g
15432018 250 mg 250mg
15422018 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15412018 Supplier Thermo Scientific Alfa Aesar Supplier No. H55694.03
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 19064-24-5 | C6H3F2NO2 | 159.092 g/mol

1,3-Difluoro-2-nitrobenzene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Difluoro-2-nitrobenzene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19064-24-5
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.092
MDL Number MFCD00192035
InChI Key SSNCMIDZGFCTST-UHFFFAOYSA-N
Synonym 2,6-difluoronitrobenzene, 1,3-difluoro-2-nitro-benzene, benzene, 1,3-difluoro-2-nitro, 2,6-difluoro nitrobenzene, pubchem4403, 1,3-difluoronitrobenzene, acmc-1buv1, 2,6-difluoro-nitrobenzene, 2,6-difluoro nitro benzene, 1,3-difluoro-2-nitrobenzen
PubChem CID 87922
IUPAC Name 1,3-difluoro-2-nitrobenzene
SMILES C1=CC(=C(C(=C1)F)[N+](=O)[O-])F

Specifications

Melting Point 0°C
Density 1.503
Boiling Point 91°C to 92°C (11 mmHg)
Flash Point 88°C (190°F)
Refractive Index 1.494
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 159.09
Percent Purity 98%
Chemical Name or Material 1,3-Difluoro-2-nitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.