missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one, 97%, Thermo Scientific Chemicals

Product Code. 10562531 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10562531

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2147-83-3
Molecular Formula C12H14N3O
Molecular Weight (g/mol) 216.26
MDL Number MFCD00044827
InChI Key YFEOSTXFQCDCAR-UHFFFAOYSA-O
Synonym 1,3-dihydro-1-1,2,3,6-tetrahydro-4-pyridinyl-2h-benzimidazole-2-one, 1,3-dihydro-1-1,2,3,6-tetrahydro-4-pyridinyl-2h-benzimidazol-2-one, 3-1,2,3,6-tetrahydropyridin-4-yl-1h-benzimidazol-2-one, 1-1,2,3,6-tetrahydro-4-pyridyl-2-benzimidazolinone, 1-1,2,3,6-tetrahydropyridin-4-yl-1,3-dihydro-2h-benzimidazol-2-one, 1-1,2,3,6-tetrahydropyridin-4-yl-2,3-dihydro-1h-1,3-benzodiazol-2-one, 1-4-1,2,5,6-tetrahydropyridyl-3-hydrobenzimidazol-2-one, 1-1,2,3,6-tetrahydropyridin-4-yl-3h-1,3-benzodiazol-2-one, benzoimidazol-2-one, 1-1,2,3,6-tetrahydro-pyridin-4-yl-1,3-dihydro
PubChem CID 75070
IUPAC Name 3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-benzimidazol-2-one
SMILES O=C1NC2=CC=CC=C2N1C1=CC[NH2+]CC1

Specifications

Melting Point 195.0°C to 200.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: 2.5g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,dmf,,dichloromethane and acetone
Formula Weight 215.25
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.