missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dihydroxyacetone dimer, 97%

Product Code. 11462257
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462257 50 g 50g
11472257 250 g 250g
11452257 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462257 Supplier Thermo Scientific Alfa Aesar Supplier No. A14189.18
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 62147-49-3 | C3H6O3 | 90.08 g/mol

1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.

Solubility
Soluble in ethanol, ethyl ether, acetone. Slightly soluble in water. Insoluble in ligroin.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62147-49-3
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00051019
InChI Key RXKJFZQQPQGTFL-UHFFFAOYSA-N
Synonym 1,3-dihydroxyacetone dimer, 1,3-dihydroxypropan-2-one dimer, 2,5-bis hydroxymethyl-1,4-dioxane-2,5-diol, 2,5-dihydroxy-1,4-dioxane-2,5-dimethanol, dihydroxyacetone dimer, 2,5-dihydroxydioxane-2,5-dimethanol, 1,4-dioxane-2,5-dimethanol, 2,5-dihydroxy, acmc-20aoav, pubchem4073
PubChem CID 4180364
SMILES OCC(=O)CO

Specifications

Melting Point ∼75°C to 82°C
Quantity 50 g
Beilstein 112910
Merck Index 14,3182
Solubility Information Soluble in ethanol,ethyl ether,acetone. Slightly soluble in water. Insoluble in ligroin.
IUPAC Name 2,5-bis(hydroxymethyl)-1,4-dioxane-2,5-diol
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material 1,3-Dihydroxyacetone dimer
compliance-icons
Product Identifier
  • 1,3-Dihydroxyacetone dimer
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1,3-Dihydroxyacetonedimer

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.