missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dihydroxyacetone dimer

Product Code. 15493339
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493339 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15493339 Supplier Thermo Scientific Alfa Aesar Supplier No. J60592.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 62147-49-3 | C3H6O3 | 90.08 g/mol

1,3-Dihydroxyacetone dimer is primarily used as an ingredient in sunless tanning products. It is a waterproof skin dye used to darken light or unpigmented areas of skin affected by vitiligo, scars or other causes.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1,3-Dihydroxyacetone dimer is primarily used as an ingredient in sunless tanning products. It is a waterproof skin dye used to darken light or unpigmented areas of skin affected by vitiligo, scars or other causes.

Solubili
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 75°C to 80°C
CAS 62147-49-3
Color White
Quantity 5 g
Physical Form Powder
Molecular Formula C3H6O3
MDL Number MFCD00051019
Merck Index 14,3182
Synonym 1,3-dihydroxyacetone dimer, 1,3-dihydroxypropan-2-one dimer, 2,5-bis hydroxymethyl-1,4-dioxane-2,5-diol, 2,5-dihydroxy-1,4-dioxane-2,5-dimethanol, dihydroxyacetone dimer, 2,5-dihydroxydioxane-2,5-dimethanol, 1,4-dioxane-2,5-dimethanol, 2,5-dihydroxy, acmc-20aoav, pubchem4073
Solubility Information Soluble in dimethyl sulfoxide,methanol,and water.
InChI Key RXKJFZQQPQGTFL-UHFFFAOYSA-N
SMILES OCC(=O)CO
IUPAC Name 2,5-bis(hydroxymethyl)-1,4-dioxane-2,5-diol
Molecular Weight (g/mol) 90.08
PubChem CID 4180364
Formula Weight 180.16
Chemical Name or Material 1,3-Dihydroxyacetone dimer
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.