missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diisopropylimidazolium chloride, 97%, ACROS Organics™

Product Code. 10413355 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10413355 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10413355 Supplier Thermo Scientific Acros Supplier No. 382430010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 139143-09-2
CAS Min % 96.0
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C9H17ClN2
MDL Number MFCD03840577
Quantity 1 g
Synonym 1,3-diisopropylimidazolium chloride, 1,3-di-i-propylimidazolium chloride, 1,3-diisopropyl-1h-imidazol-3-ium chloride, 1,3-diisopropylimidazol-1-ium chloride, diic, acmc-20ap45, n,n'-isopropyl imidazolium chloride, 1h-imidazolium,1,3-bis 1-methylethyl-, chloride 1:1, n,n inverted exclamation marka-isopropyl imidazolium chloride
InChI Key DOFXKPAOJLLPII-UHFFFAOYSA-M
SMILES [Cl-].CC(C)N1C=C[N+](=C1)C(C)C
IUPAC Name 1,3-bis(propan-2-yl)-1H-imidazol-3-ium chloride
Molecular Weight (g/mol) 188.70
PubChem CID 11229242
Formula Weight 188.7
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 1, 3-Diisopropylimidazolium chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.