missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-1H-pyrazol-5-amine, 97%, Thermo Scientific™

Product Code. 10110742
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10110742 10 g 10g
10205524 50 g 50g
10327494 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10110742 Supplier Thermo Scientific Maybridge Supplier No. KM00001EA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 3524-32-1
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.148
MDL Number MFCD00051651
InChI Key ZFDGMMZLXSFNFU-UHFFFAOYSA-N
Synonym 5-amino-1,3-dimethylpyrazole, 1,3-dimethyl-1h-pyrazol-5-amine, 1,3-dimethyl-1h-pyrazole-5yl-amine, 1h-pyrazol-5-amine, 1,3-dimethyl, 3-amino-2,5-dimethyl-2h-pyrazole, 2,5-dimethyl-2h-pyrazol-3-ylamine, 5-amino-1,3-dimethyl pyrazole, 1,3-dimethyl-1h-pyrazole-5-amine, 5-amino-1,3-dimethyl-1h-pyrazole, 1,3-dimethyl-5-aminopyrazole
PubChem CID 520721
IUPAC Name 2,5-dimethylpyrazol-3-amine
SMILES CC1=NN(C(=C1)N)C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 111.15
Percent Purity 97%
Chemical Name or Material 1, 3-Dimethyl-1H-pyrazol-5-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.