missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-1H-pyrazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10079211
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10079211 1 g 1g
10259884 5 g 5g
11350567 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10079211 Supplier Thermo Scientific Maybridge Supplier No. CC12501DA
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 5744-56-9
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD00159636
InChI Key QRWZFUBHOQWUGH-UHFFFAOYSA-N
Synonym 1,3-dimethyl-1h-pyrazole-5-carboxylic acid, 1,3-dimethylpyrazole-5-carboxylic acid, 2,5-dimethyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1,3-dimethyl, 1,3-dimethylpyrazole-5-carboxylicacid, pubchem7839, ksc269g2l, 1,3-dimethylpyrazol-5-carboxylic acid, pyrazole-5-carboxylic acid, 1,3-dmethyl, 1h-pyrazole-5-carboxylicacid, 1,3-dimethyl
PubChem CID 587721
IUPAC Name 2,5-dimethylpyrazole-3-carboxylic acid
SMILES CN1N=C(C)C=C1C(O)=O

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 1, 3-Dimethyl-1H-pyrazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.