missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-2-imidazolidinone, 98%

Product Code. 10377840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
Product Code. Quantity Packaging unitSize
10377840 100 g Glass bottle 100g
10665822 1 kg Glass bottle 1kg
2 options
This item is not returnable. View return policy

Product Code. 10377840

Brand: Thermo Scientific Acros 185931000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80-73-9 | C5H10N2O | 114.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-73-9
Molecular Formula C5H10N2O
Molecular Weight (g/mol) 114.15
MDL Number MFCD00003188
InChI Key CYSGHNMQYZDMIA-UHFFFAOYSA-N
Synonym 1,3-dimethyl-2-imidazolidinone, n,n'-dimethylethyleneurea, dimethyl imidazolidinone, n,n'-dimethylimidazolidinone, 1,3-dimethylimidazolidone, rhonite 1, dmeu, karbomos tsem, 2-imidazolidinone, 1,3-dimethyl, 1,3-dimethylethyleneurea
PubChem CID 6661
IUPAC Name 1,3-dimethylimidazolidin-2-one
SMILES CN1CCN(C)C1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 8.2°C
Density 1.0500g/mL
Boiling Point 225.5°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4710 to 1.4730
Quantity 100 g
Fieser 11,202; 15,145
Merck Index 15, 3273
Specific Gravity 1.05
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in methanol, ethanol, 2-propanol, ether,, methylene chloride, dmf, acetone, ketone and, chloroform
Viscosity 1.944 mPa.s (25°C)
Formula Weight 114.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,3-Dimethyl-2-imidazolidinone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Dimethyl-2-imidazolidinone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.