missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-2-imidazolidinone, TRC

Product Code. 30383977
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383977 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383977 Supplier Toronto Research Chemicals Supplier No. TRCD47355010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Dimethyl-2-imidazolidinone
CAS 80-73-9
Molecular Formula C5 H10 N2 O
Quantity 10 g
Synonym 2-Imidazolidinone, 1,3-dimethyl-, 1,3-Dimethyl-2-imidazolidinone, 1,3-Dimethyl-2-imidazolidindione, 1,3-Dimethyl-2-imidazolidone, 1,3-Dimethylethyleneurea, 1,3-Dimethylimidazolidinone, DMEU, DMEU (solvent), DMI, Dimethylimidazolidinone, Karbomos TsEM, N,N-Dimethylethylenediamine cyclic urea, N,N-Dimethylimidazolidinone, N,N'-Dimethyl-2-imidazolidinone, N,N'-Dimethyl-N,N'-ethyleneurea, N,N'-Dimethylethyleneurea, N,N'-Dimethylimidazolidinone, N,N'-Dimethylimidazolidone, Rhonite 1
InChI Formula InChI=1S/C5H10N2O/c1-6-3-4-7(2)5(6)8/h3-4H2,1-2H3
SMILES CN1CCN(C)C1=O
IUPAC Name 1,3-dimethylimidazolidin-2-one
Molecular Weight (g/mol) 114.146
Formula Weight 114.0793
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.