missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-2-nitrobenzene, TRC

Product Code. 30387625
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387625 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387625 Supplier Toronto Research Chemicals Supplier No. TRCD4764451G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 81-20-9
Molecular Formula C8 H9 N O2
Quantity 1 g
Synonym m-Xylene, 2-nitro- (6CI,7CI,8CI), 1,3-Dimethyl-2-nitrobenzene, 2,6-Dimethylnitrobenzene, 2-Nitro-1,3-dimethylbenzene, 2-Nitro-1,3-xylene, 2-Nitro-m-xylene, NSC 9811
InChI Formula InChI=1S/C8H9NO2/c1-6-4-3-5-7(2)8(6)9(10)11/h3-5H,1-2H3
SMILES Cc1cccc(C)c1[N+](=O)[O-]
IUPAC Name 1,3-dimethyl-2-nitrobenzene
Molecular Weight (g/mol) 151.16
Formula Weight 151.0633
Chemical Name or Material 1,3-Dimethyl-2-nitrobenzene (2-Nitro-m-xylene)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.