missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone, 97%, pure

Product Code. 10543201 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10543201 500 mL Glass bottle 500mL
10003000 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10543201 Supplier Thermo Scientific Acros Supplier No. 224645000
  • ONLINE EXCLUSIVE PRICE
    195.00 €
    179.00 € / 500mL
    Save 16.00 €
    8% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7226-23-5 | C6H12N2O | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7226-23-5
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.17
MDL Number MFCD00006550
InChI Key GUVUOGQBMYCBQP-UHFFFAOYSA-N
Synonym 1,3-dimethyl-3,4,5,6-tetrahydro-2 1h-pyrimidinone, 1,3-dimethyltetrahydropyrimidin-2 1h-one, dmpu, dimethylpropyleneurea, 2 1h-pyrimidinone, tetrahydro-1,3-dimethyl, n,n'-dimethylpropyleneurea, n,n'-dimethyltrimethyleneurea, ccris 4322, n,n'-dimethylpropylene urea, tetrahydro-1,3-dimethyl-2 1h pyrimidine
PubChem CID 81646
IUPAC Name 1,3-dimethyl-1,3-diazinan-2-one
SMILES CN1CCCN(C1=O)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -20°C
Color Colorless to Yellow
Density 1.0640g/mL
Boiling Point 246°C
Flash Point 121°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4875 to 1.4895
Quantity 500 mL
Beilstein 24, IV, 32
Fieser 11,207; 13,122; 16,146
Specific Gravity 1.064
Solubility Information Solubility in water: freely soluble
Viscosity 3.41 mPa.s (20°C)
Formula Weight 128.17
Percent Purity 97%
Grade Pure
Physical Form Liquid
Chemical Name or Material 1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H361f-Suspected of damaging fertility.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.