missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyladamantane, TRC

Product Code. 30007075
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007075 25 g 25g
30004918 1 g 1g
30004899 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007075 Supplier Toronto Research Chemicals Supplier No. TRCD46077825G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 702-79-4
Formula Weight 164.1565
InChI Formula InChI=1S/C12H20/c1-11-4-9-3-10(5-11)7-12(2,6-9)8-11/h9-10H,3-8H2,1-2H3
IUPAC Name 1,3-dimethyladamantane
Molecular Formula C12 H20
Molecular Weight (g/mol) 164.29
SMILES CC12CC3CC(CC(C)(C3)C1)C2
Synonym 1,3-Dimethyladamantane, Memantine Related Compound A (USP), Tricyclo[3.3.1.13,7]decane, 1,3-dimethyl-, Adamantane, 1,3-dimethyl- (6CI,8CI), 1,3-Dimethyltricyclo[3.3.1.13,7]decane, 3,5-Dimethyladamantane
Chemical Name or Material 1,3-Dimethyladamantane

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.