missing translation for 'onlineSavingsMsg'
Learn More

1-[3-(Dimethylamino)propyl]-3-ethylurea, TRC

Product Code. 30375122
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30375122 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375122 Supplier Toronto Research Chemicals Supplier No. TRCD492553250MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[3-(Dimethylamino)propyl]-3-ethylurea
CAS 32897-26-0
Molecular Formula C8H19N3O
Quantity 250 mg
Synonym N-[3-(Dimethylamino)propyl]-N'-ethylurea, 1-Ethyl-3-(3-dimethylaminopropyl)urea, EDC Urea
InChI Formula InChI=1S/C8H19N3O/c1-4-9-8(12)10-6-5-7-11(2)3/h4-7H2,1-3H3,(H2,9,10,12)
SMILES CCNC(=O)NCCCN(C)C
IUPAC Name 1-[3-(dimethylamino)propyl]-3-ethylurea
Molecular Weight (g/mol) 173.26
Formula Weight 173.1528
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.