missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 97%

Product Code. 15169293 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15169293 250 g Glass bottle 250g
10210120 10 g Glass bottle 10g
11955591 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15169293 Supplier Thermo Scientific Acros Supplier No. 438842500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1892-57-5 | C8H17N3 | 155.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1892-57-5
Molecular Formula C8H17N3
Molecular Weight (g/mol) 155.24
InChI Key LMDZBCPBFSXMTL-UHFFFAOYSA-N
Synonym 1-3-dimethylaminopropyl-3-ethylcarbodiimide, edac, 1-ethyl-3-3-dimethylaminopropyl carbodiimide, n1-ethylimino methylene-n3,n3-dimethylpropane-1,3-diamine, unii-rj5ozg6i4a, edci, 1,3-propanediamine, n'-ethylcarbonimidoyl-n,n-dimethyl, n-3-dimethylaminopropyl-n'-ethylcarbodiimide, rj5ozg6i4a, ethyldimethylaminopropyl carbodiimide
PubChem CID 15908
IUPAC Name 3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine
SMILES CCN=C=NCCCN(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8850g/mL
Boiling Point 66.0°C to 68.0°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4600 to 1.4620 (20°C, 589nm)
Quantity 250 g
Merck Index 15,3555
Specific Gravity 0.885
Formula Weight 155.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide
Show More Show Less
compliance-icons
Product Identifier
  • 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.