missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%

Product Code. 10397490 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
10g
1g
500g
50g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10397490 10 g Glass Bottle 10g
10225750 1 g Glass Bottle 1g
10078001 50 g Glass Bottle 50g
10363712 100 g Glass Bottle 100g
10308243 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10397490 Supplier Thermo Scientific Acros Supplier No. 171440100
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25952-53-8 | C8H18ClN3 | 191.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25952-53-8
Molecular Formula C8H18ClN3
Molecular Weight (g/mol) 191.70
MDL Number MFCD00012503
InChI Key FPQQSJJWHUJYPU-UHFFFAOYSA-N
Synonym 1-3-dimethylaminopropyl-3-ethylcarbodiimide hydrochloride, edc.hcl, edci, edc hydrochloride, wsc hcl, edac hydrochloride, edac, hydrochloride, 1-ethyl-3-3-dimethylaminopropyl carbodiimide hydrochloride, edcl, edac hcl
PubChem CID 2723939
IUPAC Name 3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES Cl.CCN=C=NCCCN(C)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 111.0°C to 116.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (Argentometry)
Packaging Glass Bottle
Linear Formula (CH3)2NCH2CH2CH2N=C=NCH2CH3
Quantity 10 g
UN Number 2811
Fieser 12,199
Merck Index 15, 3555
Solubility Information Solubility in water: >200g/l. Other solubilities: soluble in ethanol
Formula Weight 191.71
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide Hydrochloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.