missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%

Product Code. 11404393
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11404393

Brand: Thermo Scientific Alfa Aesar A10962.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22572-40-3 | C9H20N3+ | 170.28 g/mol

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide is in proteins as water soluble carboxyl modifying reagent. It is used as a crosslinking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide is in proteins as water soluble carboxyl modifying reagent. It is used as a crosslinking agent.

Solubility
Soluble in water.

Notes
Keep in a dry, cool and well-ventilated place. Keep container tightly closed. Incompatible with acids, strong oxidizing agents, moist air and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22572-40-3
Molecular Formula C9H20N3+
Molecular Weight (g/mol) 170.28
MDL Number MFCD00011833
InChI Key JHMDMCSGQQLHNA-UHFFFAOYSA-N
Synonym 3-ethylimino methylidene amino-n,n,n-trimethylpropan-1-aminium, 1-3-trimethylaminio propyl-3-ethylcarbodiimide, 3-ethyliminomethylideneamino propyl-trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide methiodide, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 3-ethylimino methylidene amino propyl trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 200-400 mesh, 2 % cross-linked with divinylbenzene
PubChem CID 122243
IUPAC Name 3-(ethyliminomethylideneamino)propyl-trimethylazanium
SMILES CCN=C=NCCC[N+](C)(C)C

Specifications

Melting Point 90°C to 92°C
Quantity 1 g
Beilstein 4162760
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in water.
Formula Weight 297.17
Percent Purity 98%
Chemical Name or Material 1- (3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.