missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%

Product Code. p-115293 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10798162

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22572-40-3 | C9H20IN3 | 297.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22572-40-3
Molecular Formula C9H20IN3
Molecular Weight (g/mol) 297.18
MDL Number MFCD00011833
InChI Key JHMDMCSGQQLHNA-UHFFFAOYSA-N
Synonym 3-ethylimino methylidene amino-n,n,n-trimethylpropan-1-aminium, 1-3-trimethylaminio propyl-3-ethylcarbodiimide, 3-ethyliminomethylideneamino propyl-trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide methiodide, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 3-ethylimino methylidene amino propyl trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 200-400 mesh, 2 % cross-linked with divinylbenzene
PubChem CID 122243
IUPAC Name 3-(ethyliminomethylideneamino)propyl-trimethylazanium
SMILES CCN=C=NCCC[N+](C)(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 92.0°C to 97.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 3555
Solubility Information Solubility in water: 50g/L
Formula Weight 297.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.