missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethylurea, 98%

Product Code. p-120250 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
1kg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10723651 5 g Glass Bottle 5g
10686872 100 g Plastic bottle 100g
10686682 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10723651 Supplier Thermo Scientific Acros Supplier No. 116910050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 96-31-1 | C3H8N2O | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-31-1
Molecular Formula C3H8N2O
Molecular Weight (g/mol) 88.11
MDL Number MFCD00008286
InChI Key MGJKQDOBUOMPEZ-UHFFFAOYSA-N
Synonym n,n'-dimethylurea, sym-dimethylurea, urea, n,n'-dimethyl, dimethylurea, symmetric dimethylurea, urea, 1,3-dimethyl, n,n'-dimethylharnstoff, 1,3-dimethyl urea, n,n'-dimethylharnstoff german, unii-wam6dr9i4x
PubChem CID 7293
ChEBI CHEBI:80472
IUPAC Name 1,3-dimethylurea
SMILES CNC(=O)NC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 101°C to 105°C
Color White
Boiling Point 268°C to 270°C
Flash Point 157°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula CH3NHCONHCH3
Quantity 5 g
Beilstein 04, 65
Solubility Information Solubility in water: 765g/L in water (21.5°C). Other solubilities: insoluble in ether,soluble in alcohols
Formula Weight 88.11
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material 1, 3-Dimethylurea, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.