missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxolan-2-one, TRC

Product Code. 30009401
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009401 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30009401 Supplier Toronto Research Chemicals Supplier No. TRCD4865255G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 96-49-1
Molecular Formula C3 H4 O3
Quantity 5 g
Synonym Carbonic acid, cyclic ethylene ester (6CI,8CI), 2-Dioxolanone, 2-Oxo-1,3-dioxolane, Cyclic ethylene carbonate, Ethylene carbonate, Ethylene glycol carbonate, Glycol carbonate, JS 1002, Jeffsol EC, NSC 11801, NSC 16568, PC Dilanon, Texacar EC, 1,3-Dioxolan-2-one
InChI Formula InChI=1S/C3H4O3/c4-3-5-1-2-6-3/h1-2H2
SMILES O=C1OCCO1
IUPAC Name 1,3-dioxolan-2-one
Molecular Weight (g/mol) 88.06
Formula Weight 88.016
Chemical Name or Material 1,3-Dioxolan-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.