missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxolane, 99.5%, stab.

Product Code. 11350148
Change view
Click to view available options
Quantity:
250 mL
1 L
4 x 1 L
Conditionnement:
1 L
250mL
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
11350148 250 mL 250mL
11330148 1 L 1 L
11340148 4 x 1 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11350148 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur 042897.AK
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

As a solvent, in Mannich reaction, as an electrolyte in batteries | CAS: 646-06-0 | C3H6O2 | 74.079 g/mol

As a solvent, in Mannich reaction, as an electrolyte in batteries.1,3-Dioxolane is used as a solvent for paints, cellulose esters and in organic synthesis. It is used as an extragent for waxes, fats and oils, stabilizer for halogenated hydrocarbons and co monomer in polyacetals preparation. It is used as finishing agent and swelling agent in textiles. It acts as an intermediate for the preparation of acyclovir and vandetanib. It is an essential ingredient in industrial polymers. In polymerization reactions, it acts as a chain length regulator and a chain transfer agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 646-06-0
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00003207
InChI Key WNXJIVFYUVYPPR-UHFFFAOYSA-N
Synonym dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech
PubChem CID 12586
ChEBI CHEBI:87597
IUPAC Name 1,3-dioxolane
SMILES C1COCO1

Spécification

Melting Point -95°C
Density 1.06 g/mL
Boiling Point 74°C to 75°C
Flash Point 1°C (34°F)
Refractive Index 1.4
Quantity 250 mL
UN Number UN1166
Solubility Information Miscible with water,ethanol,ether,n-octanol,acetone,chloroform,dichloromethane,ethyl Acetate and methanol.
Formula Weight 74.08
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material 1,3-Dioxolane, Stabilized
Afficher plus Afficher moins
compliance-icons compliance-icons
Identifiant produit
  • 1,3-Dioxolane
Mention d'avertissement
  • Danger
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Mention de danger
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Informations supplémentaires
  • MIXTURE LIST-Contain: 1,3-Dioxolane

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.