missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxolane (Stabilized with BHT), TRC

Product Code. 30375209
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375209 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375209 Supplier Toronto Research Chemicals Supplier No. TRCD486800500ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Dioxolane
CAS 646-06-0
Molecular Formula C3 H6 O2
Quantity 500 mL
Synonym Dioxolane (7CI), 1,3-Dioxacyclopentane, 1,3-Dioxolan, 1,3-Dioxole, dihydro-, 5-Crown-2, DIOX, Ethylene glycol formal, Formal glycol, Glycolformal, 1,3-Dioxolane
InChI Formula InChI=1S/C3H6O2/c1-2-5-3-4-1/h1-3H2
SMILES C1COCO1
IUPAC Name 1,3-dioxolane
Molecular Weight (g/mol) 74.08
Formula Weight 74.0368
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.