missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenylisobenzofuran, 97%

Product Code. 10091620 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10091620

Brand: Thermo Scientific Acros 147580050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5471-63-6 | C20H14O | 270.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5471-63-6
Molecular Formula C20H14O
Molecular Weight (g/mol) 270.32
MDL Number MFCD00005931
InChI Key ZKSVYBRJSMBDMV-UHFFFAOYSA-N
Synonym 1,3-diphenylisobenzofuran, diphenylisobenzofuran, isobenzofuran, 1,3-diphenyl, 1,3 diphenylisobenzofuran, 2,5-diphenyl-3,4-benzofuran, acmc-209lj9, 1,3-diphenylbenzo c furan, 1,3-diphenylisobenzo b furan, 1,3-di phenyl-2-benzofuran, #
PubChem CID 21649
IUPAC Name 1,3-diphenyl-2-benzofuran
SMILES C1=CC=C(C=C1)C2=C3C=CC=CC3=C(O2)C4=CC=CC=C4

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 129.0°C to 133.0°C
Color Yellow to Green
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 17, 87
Fieser 01,342; 02,178; 04,209
Formula Weight 270.32
Percent Purity 97%
Physical Form Crystalline Solid or Needles
Chemical Name or Material 1, 3-Diphenylisobenzofuran, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.