missing translation for 'onlineSavingsMsg'
Learn More

1,3-Indanedione, 97%

Product Code. 11914851 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11914851 100 g Glass bottle 100g
10366850 5 g Glass bottle 5g
10376850 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11914851 Supplier Thermo Scientific Acros Supplier No. 122091000
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 606-23-5 | C9H6O2 | 146.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-23-5
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.15
InChI Key UHKAJLSKXBADFT-UHFFFAOYSA-N
Synonym 1,3-indanedione, 1,3-indandione, 1h-indene-1,3 2h-dione, 1,3-diketohydrindene, indan-1,3-dione, 1,3-indanone, indane-1,3-dione, diketohydrindene, 1,3-indandion, unii-4djn7yg35g
PubChem CID 11815
ChEBI CHEBI:78877
IUPAC Name indene-1,3-dione
SMILES C1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 129.0°C to 132.0°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 g
Beilstein 07,694
Formula Weight 146.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1, 3-Indanedione
compliance-icons
Product Identifier
  • 1,3-Indanedione
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.